Skip to Content
MilliporeSigma
All Photos(1)

Documents

530824

Sigma-Aldrich

2-Benzylphenyl isocyanate

97%

Synonym(s):

2′-Benzylphenyl isocyanate, 2′-Isocyanatodiphenylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2C6H4NCO
CAS Number:
Molecular Weight:
209.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.591 (lit.)

bp

309 °C (lit.)

density

1.11 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C=Nc1ccccc1Cc2ccccc2

InChI

1S/C14H11NO/c16-11-15-14-9-5-4-8-13(14)10-12-6-2-1-3-7-12/h1-9H,10H2

InChI key

GLYMYAJKKMZFQT-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service