Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

530611

Sigma-Aldrich

3-Methylbenzofuran-2-carboxylic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O3
CAS Number:
Molecular Weight:
176.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

194-197 °C (lit.)

SMILES string

Cc1c(oc2ccccc12)C(O)=O

InChI

1S/C10H8O3/c1-6-7-4-2-3-5-8(7)13-9(6)10(11)12/h2-5H,1H3,(H,11,12)

InChI key

YMZTUCZCQMQFMK-UHFFFAOYSA-N

General description

3-Methylbenzofuran-2-carboxylic acid is a benzofuran derivative. It undergoes palladium-catalyzed cross-coupling reaction with 4-iodoanisole and diphenyliodonium triflate under different reaction conditions to form the corresponding biaryl.

Application

3-Methylbenzofuran-2-carboxylic acid may be used in the preparation of 3-methyl-N-phenylbenzofuran-2-carboxamide by reacting with aniline.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karla-Sue C Marriott et al.
Bioorganic & medicinal chemistry, 20(23), 6856-6861 (2012-10-23)
Novel benzofuran-2-carboxamide ligands, which are selective for sigma receptors, have been synthesized via a microwave-assisted Perkin rearrangement reaction and a modified Finkelstein halogen-exchange used to facilitate N-alkylation. The ligands synthesized are the 3-methyl-N-phenyl-N-(3-(piperidin-1-yl)propyl)benzofuran-2-carboxamides (KSCM-1, KSCM-5 and KSCM-11). The benzofuran-2-carboxamide structure
Synthesis of biaryls via decarboxylative Pd-catalyzed cross-coupling reaction.
Becht JM, et al.
Organic Letters, 9(9), 1781-1783 (2007)
Biaryl Synthesis via Decarboxylative Pd-Catalyzed Reactions of Arenecarboxylic Acids and Diaryliodonium Triflates.
Becht JM and Drian CL.
Organic Letters, 10(14), 3161-3164 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service