Skip to Content
MilliporeSigma
All Photos(2)

Documents

529990

Sigma-Aldrich

4-Bromomethyl benzoyl bromide

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2C6H4C(O)Br
CAS Number:
Molecular Weight:
277.94
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

bp

100-112 °C/0.5 mmHg (lit.)

mp

57-61 °C (lit.)

SMILES string

BrCc1ccc(cc1)C(Br)=O

InChI

1S/C8H6Br2O/c9-5-6-1-3-7(4-2-6)8(10)11/h1-4H,5H2

InChI key

VUYGJYAPDGKPBF-UHFFFAOYSA-N

Related Categories

General description

4-Bromomethyl benzoyl bromide is a para-substituted benzoyl bromide.

Application

4-Bromomethyl benzoyl bromide may be used in the preparation of 4-(bromomethyl)-N-[(S)-3-ethyl-3-hydroxy-1-phenylpentan-2-yl]benzamide and α-methoxy-ω-4-(bromomethyl) benzoic acid ester-poly(ethylene glycol) 2000.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Systematic MALDI-TOF CID investigation on different substituted mPEG 2000.
Knop K, et al.
Macromolecular Chemistry and Physics, 211(6), 677-684 (2010)
Enantioselective addition of phenylacetylene to aldehydes catalyzed by silica-immobilized titanium (IV) complex of ?-hydroxyamide.
Huang LN, et al.
J. Mol. Catal. A: Chem., 275(1), 9-13 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service