Skip to Content
MilliporeSigma
All Photos(1)

Documents

52961

Sigma-Aldrich

4,4,5,5,6,6,7,7,7-Nonafluoroheptyl iodide

≥97.0% (GC)

Synonym(s):

1,1,1,2,2,3,3,4,4-Nonafluoro-7-iodoheptane, 1H,1H,2H,2H,3H,3H-Perfluoroheptyl iodide, 3-(Perfluorobutyl)propyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6F9I
CAS Number:
Molecular Weight:
388.01
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥97.0% (GC)

refractive index

n20/D 1.381

density

1.828 g/mL at 25 °C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)CCCI

InChI

1S/C7H6F9I/c8-4(9,2-1-3-17)5(10,11)6(12,13)7(14,15)16/h1-3H2

InChI key

BPQUIZZKLVTZOY-UHFFFAOYSA-N

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

170.1 °F

Flash Point(C)

76.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunfang Liu et al.
Cell discovery, 6, 15-15 (2020-03-29)
Liver metastasis, characterized by the spread of tumors to the liver from other areas, represents a deadly disease with poor prognosis. Currently, there is no effective therapeutic strategies and/or agents to combat liver metastasis primarily due to the insufficient understanding
Shira Yomtoubian et al.
Cell reports, 30(3), 755-770 (2020-01-23)
Epigenetic changes are increasingly being appreciated as key events in breast cancer progression. However, breast cancer subtype-specific epigenetic regulation remains poorly investigated. Here we report that EZH2 is a leading candidate of epigenetic modulators associated with the TNBC subtype and
Shengchen Lin et al.
Cell reports, 28(11), 2824-2836 (2019-09-12)
The deregulation of the actin cytoskeleton has been extensively studied in metastatic dissemination. However, the post-dissemination role of the actin cytoskeleton dysregulation is poorly understood. Here, we report that fascin, an actin-bundling protein, promotes lung cancer metastatic colonization by augmenting
Yosuke Funato et al.
Developmental cell, 55(4), 387-397 (2020-09-13)
Extracellular pH is usually maintained around 7.4 in multicellular organisms, and cells are optimized to proliferate under this condition. Here, we find cells can adapt to a more acidic pH of 6.5 and become addicted to this acidic microenvironment by
Zhe Zhu et al.
Cell stem cell, 26(2), 187-204 (2020-01-21)
Zika virus (ZIKV) causes microcephaly by killing neural precursor cells (NPCs) and other brain cells. ZIKV also displays therapeutic oncolytic activity against glioblastoma (GBM) stem cells (GSCs). Here we demonstrate that ZIKV preferentially infected and killed GSCs and stem-like cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service