Skip to Content
MilliporeSigma
All Photos(1)

Documents

52927

Sigma-Aldrich

(S)-3-(Boc-amino)pyrrolidine

≥98.0% (TLC)

Synonym(s):

tert-Butyl (S)-3-pyrrolidinylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N2O2
CAS Number:
Molecular Weight:
186.25
Beilstein:
5377813
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (TLC)

form

solid

optical activity

[α]/D -21.5±2.0°, c = 1 in ethanol

SMILES string

CC(C)(C)OC(=O)N[C@H]1CCNC1

InChI

1S/C9H18N2O2/c1-9(2,3)13-8(12)11-7-4-5-10-6-7/h7,10H,4-6H2,1-3H3,(H,11,12)/t7-/m0/s1

InChI key

DQQJBEAXSOOCPG-ZETCQYMHSA-N

Application

(S)-3-(Boc-amino)pyrrolidine can be used as a building block to prepare:
  • 2,4,6-trisubstitued pyrido[3,4-d]pyrimidine derivatives as potent inhibitors against EGFR tyrosine kinase.
  • Aminopyrrolidine scaffolds for asymmetric Morita−Baylis-Hillman reaction.
  • N-benzyl-3-sulfonamidopyrrolidines as potent bacterial cell division inhibitors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and biological evaluation of irreversible EGFR tyrosine kinase inhibitors containing pyrido [3, 4-d] pyrimidine scaffold
Zhang H, et al.
Bioorganic & Medicinal Chemistry, 26(12), 3619-3633 (2018)
Chiral 3-aminopyrrolidines as a rigid diamino scaffold for organocatalysis and organometallic chemistry
Pouliquen M, et al.
Tetrahedron Asymmetry, 21(11-12), 1511-1521 (2010)
N-Benzyl-3-sulfonamidopyrrolidines as novel inhibitors of cell division in E. coli
Mukherjee S, et al.
Bioorganic & medicinal chemistry letters, 17(23), 6651-6655 (2007)

Articles

Mono-Boc-protected diamines are versatile building blocks for chemical synthesis. Their production is a lot more challenging than the simple reaction scheme might imply, because the Boc-anhydride reagent cannot differentiate between the two identical amino moieties in the substrate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service