Skip to Content
MilliporeSigma
All Photos(1)

Documents

529028

Sigma-Aldrich

Ethyl 5-Bromo-2-chlorobenzoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(Cl)CO2C2H5
CAS Number:
Molecular Weight:
263.52
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.56 (lit.)

bp

288 °C (lit.)

density

1.55 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)c1cc(Br)ccc1Cl

InChI

1S/C9H8BrClO2/c1-2-13-9(12)7-5-6(10)3-4-8(7)11/h3-5H,2H2,1H3

InChI key

AMGWDYLEMSMUIO-UHFFFAOYSA-N

Application

Ethyl 5-Bromo-2-chlorobenzoate may be used in the synthesis of ethyl 6-bromo-3-chloro-3′-methylbiphenyl-2-carboxylate.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New mixed Li/Mg and Li/Mg/Zn amides for the chemoselective metallation of arenes and heteroarenes.
Rohbogner CJ, et al.
European Journal of Organic Chemistry, 11, 1781-1795 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service