Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

528897

Sigma-Aldrich

5-Bromo-2-iodotoluene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(Br)I
CAS Number:
Molecular Weight:
296.93
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.65 (lit.)

bp

263 °C (lit.)

density

2.08 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(Br)ccc1I

InChI

1S/C7H6BrI/c1-5-4-6(8)2-3-7(5)9/h2-4H,1H3

InChI key

GHTUADBHTFHMNI-UHFFFAOYSA-N

General description

5-Bromo-2-iodotoluene is a halogenated hydrocarbon. It undergoes chemoselective Suzuki reaction with phenylboronic acid to yield the corresponding carboxylic acid.[1]

Application

5-Bromo-2-iodotoluene may be used in the synthesis of:
  • 2-bromo-5-(4-bromo-2-methylphenyl)-3-methylpyridine[2]
  • 4-bromo-4′-(carbazol-9-yl)-2-methylbiphenyl[3]
  • 4,4′-dibromo-2-methyl-biphenyl[4]
  • 3-methyl-4-phenylbromobenzene[1]
  • 4-(4-bromo-2-methylphenyl)butanoic acid[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Jonathan Fray et al.
Journal of medicinal chemistry, 46(16), 3514-3525 (2003-07-25)
The pathology of chronic dermal ulcers is characterized by excessive proteolytic activity which degrades extracellular matrix (required for cell migration) and growth factors and their receptors. The overexpression of MMP-3 (stromelysin-1) and MMP-13 (collagenase-3) is associated with nonhealing wounds, whereas
Synthesis of comb polyphenylenes by Suzuki coupling from AB macromonomers.
Zhou S, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 52(11), 1519-1524 (2014)
Synthesis, Properties and Applications of Biphenyl Functionalized 9, 9-Bis (4-diphenylaminophenyl) fluorenes as Bifunctional Materials for Organic Electroluminescent Devices.
Thangthong A-M, et al.
European Journal of Organic Chemistry, 27, 5263-5274 (2012)
Synthesis of new phenylpyridyl scaffolds using the Garlanding approach.
Voisin-Chiret AS, et al.
Tetrahedron, 66(40), 8000-8005 (2010)
E Scott Priestley et al.
Journal of medicinal chemistry, 58(15), 6225-6236 (2015-07-08)
On the basis of a crystal structure of a phenylpyrrolidine lead and subsequent molecular modeling results, we designed and synthesized a novel series of macrocyclic FVIIa inhibitors. The optimal 16-membered macrocycle was 60-fold more potent than an acyclic analog. Further

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service