Skip to Content
MilliporeSigma
All Photos(1)

Documents

528595

Sigma-Aldrich

2,6-Diethoxybenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2C6H3CO2H
CAS Number:
Molecular Weight:
210.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

131-135 °C (lit.)

SMILES string

CCOc1cccc(OCC)c1C(O)=O

InChI

1S/C11H14O4/c1-3-14-8-6-5-7-9(15-4-2)10(8)11(12)13/h5-7H,3-4H2,1-2H3,(H,12,13)

InChI key

XUIVSLVJEQVKLB-UHFFFAOYSA-N

General description

2,6-Diethoxybenzoic acid undergoes catalytic hydrodecarboxylation in the presence of toluene/D2O to afford 2-d-1,3-diethoxybenzene.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Zhang et al.
Journal of the American Chemical Society, 135(46), 17270-17273 (2013-11-01)
Through-space metal/hydrogen shift is an important strategy for transition-metal-catalyzed C-H bond activation. Here we describe the synthesis and characterization of a Rh(I) 2,6-dimethoxybenzoate complex that underwent stoichiometric rearrangement via a highly unusual 1,3-rhodium migration. This aryl-to-aryl 1,3-Rh/H shift was also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service