Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

527742

Sigma-Aldrich

2-Cyanobenzoic acid

technical grade

Synonym(s):

o-Carboxybenzonitrile, NSC 193443

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4CO2H
CAS Number:
Molecular Weight:
147.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

solid

mp

212 °C (lit.)

functional group

carboxylic acid
nitrile

SMILES string

OC(=O)c1ccccc1C#N

InChI

1S/C8H5NO2/c9-5-6-3-1-2-4-7(6)8(10)11/h1-4H,(H,10,11)

InChI key

DTNSDCJFTHMDAK-UHFFFAOYSA-N

General description

2-Cyanobenzoic acid can be prepared from phthalic anhydride.[1] Mass spectrophotometry, NMR, IR are extremely useful techniques to determine the ring-chain tautomeric equilibrium of 2-cyanobenzoic acid in all three phases.[2] The standard (p° = 0.1MPa) molar enthalpy of formation of 2-cyanobenzoic acid is -(150.7 ± 2.0) kJ.mol-1.[3]

Application

2-Cyanobenzoic acid may be used to synthesize phthalamidine.[4]
Reactant involved in the synthesis of:
  • T-box riboswitch antiterminator RNA-binding oxazolidinone derivatives
  • Benzoate modified β-cyclodextrin derivatives
  • Pyrrolo[1,2-f]triazines for use as JAK2 inhibitors

Reactant involved in:
  • Decarboxylation
  • Decarboxylative halogenation
  • Protodecarboxylation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Experimental Study of Tautomeric Equilibria of 2-Cyanobenzoic Acids in Gas, Solution and Solid Phase.
Iglesias DA, et al.
Journal of Applied Solution Chemistry and Modeling, 1(2), 113-113 (2012)
The Study of Novel Synthetic Method of 2-Cyanobenzo.
Yang YS, et al.
Guangzhou Chemical Industry/ Guangzhou Huagong, 22, 016-016 (2011)
Standard molar enthalpies of formation of 2-, 3-and 4-cyanobenzoic acids.
da Silva MAVR, et al.
The Journal of Chemical Thermodynamics, 40(8), 1226-1231 (2008)
Studies on the syntheses of heterocyclic compounds. CCIV. Ring contraction in the catalytic hydrogenation of 3-oximino-4-oxoisocarbostyril.
Kametani T, et al.
Chemical & Pharmaceutical Bulletin, 15(12), 1916-1921 (1967)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service