Skip to Content
MilliporeSigma
All Photos(1)

Documents

527718

Sigma-Aldrich

2-Ethylbenzonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4CN
CAS Number:
Molecular Weight:
131.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.523 (lit.)

bp

94-95 °C/12 mmHg (lit.)

density

0.974 g/mL at 25 °C (lit.)

SMILES string

CCc1ccccc1C#N

InChI

1S/C9H9N/c1-2-8-5-3-4-6-9(8)7-10/h3-6H,2H2,1H3

InChI key

UZDXATQPJOOHQJ-UHFFFAOYSA-N

General description

2-Ethylbenzonitrile is present as one of the constituents in the bioactive neutral sub-fraction of cigarette smoke condensate. It can be obtained from 2-ethylaniline, via Sandmeyer reaction. Density and the refractive index of 2-ethylbenzonitrile at 25°C are 0.9560g/cm3 and 1.5270, respectively.

Application

2-Ethylbenzonitrile may be used to synthesize 2-ethylhexahydrobenzylamine and 2-ethylstyrene.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
The Yaws Handbook of Vapor Pressure: Antoine coefficients , 234-234 (2015)
Alkylphenols and arylnitriles in a biologically active neutral subfraction of cigarette smoke condensate.
Miller RL and Stedman RL.
Phytochemistry, 10(5), 1135-1140 (1971)
Arrhenius parameters for rearrangements of the neophyl, 1-indanylmethyl, 2-allylbenzyl, and 2-(2-vinylphenyl) ethyl radicals relative to hydrogen abstraction from tributylstannane.
Franz JA, et al.
Journal of the American Chemical Society, 106(14), 3964-3967 (1984)
1-Cyano-1, 3-butadienes. II. Carbon Structure of the Adduct Formed by the Diels--Alder Condensation of 1-Cyano-1, 3-butadiene with 1, 3-Butadiene.
Snyder HR and Poos GI.
Journal of the American Chemical Society, 71(3), 1057-1058 (1949)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service