Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

527246

Sigma-Aldrich

N-(tert-Butoxycarbonyl)-4-bromoaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4NHCO2C(CH3)3
CAS Number:
Molecular Weight:
272.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

103-106 °C (lit.)

SMILES string

CC(C)(C)OC(=O)Nc1ccc(Br)cc1

InChI

1S/C11H14BrNO2/c1-11(2,3)15-10(14)13-9-6-4-8(12)5-7-9/h4-7H,1-3H3,(H,13,14)

InChI key

VLGPDTPSKUUHKR-UHFFFAOYSA-N

General description

N-(tert-Butoxycarbonyl)-4-bromoaniline is an N-(tert-butyloxycarbonyl) [N-BOC] protected 4-bromo aniline. Due to the presence of BOC protection, it serves as an ideal substrate for Suzuki coupling reactions. It affords biaryls via reaction with pyridine or 3-benzaldehyde boronic acids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coutts IGC, et al.
Tetrahedron Letters, 38, 5563-5563 (1997)
M Lamothe et al.
Journal of medicinal chemistry, 40(22), 3542-3550 (1997-11-14)
The synthesis and binding affinity at cloned h5-HT1D, h5-HT1D, and h5-HT1A receptors of 3-[3-(N,N-dimethylamino)propyl]-4-hydroxy- N-[4-(pyridin-4-yl)phenyl]benzamide (2, GR-55562) and four O-methylated analogs are described. The functional activity of these compounds was determined at the h5-HT1B receptor using a [35S]GTP gamma S

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service