Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

526916

Sigma-Aldrich

4-Fluoro-3-nitrobenzyl alcohol

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(NO2)CH2OH
CAS Number:
Molecular Weight:
171.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

96%

mp

39-44 °C (lit.)

SMILES string

OCc1ccc(F)c(c1)[N+]([O-])=O

InChI

1S/C7H6FNO3/c8-6-2-1-5(4-10)3-7(6)9(11)12/h1-3,10H,4H2

InChI key

MKWJZTFMDWSRIH-UHFFFAOYSA-N

General description

4-Fluoro-3-nitrobenzyl alcohol can be prepared from the reaction between borane (in THF) and 4-fluoro-3-nitrobenzoic acid (in THF) at 0°C.

Application

4-Fluoro-3-nitrobenzyl alcohol may be used in the synthesis of 4-fluoro-3-nitrobenzyl chloride.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Experimentally induced phenylketonuria. II. Potential inhibitors of phenylalanine hydroxylase.
J I DeGraw et al.
Journal of medicinal chemistry, 11(2), 225-227 (1968-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service