Skip to Content
MilliporeSigma
All Photos(1)

Documents

526444

Sigma-Aldrich

2,4,5-Trimethylbenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2CO2H
CAS Number:
Molecular Weight:
164.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

150-153 °C (lit.)

SMILES string

Cc1cc(C)c(cc1C)C(O)=O

InChI

1S/C10H12O2/c1-6-4-8(3)9(10(11)12)5-7(6)2/h4-5H,1-3H3,(H,11,12)

InChI key

QENJZWZWAWWESF-UHFFFAOYSA-N

General description

Crystal structure of 2,4,5-trimethylbenzoic acid (2,4,5-TMBA) has been determined by X-ray studies. Two molecules of 2,4,5-TMBA on association affords centrosymmetric carboxyl dimers. 2,4,5-TMBA can be synthesized from pseudocumene. Vibrational analysis of monomeric and dimeric foms of 2,4,5-TMBA have been reported.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrazines and related compounds; a new synthesis of hydroxypyrazines.
R G JONES
Journal of the American Chemical Society, 71(1), 78-81 (1949-01-01)
Mehmet Karabacak et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 134, 598-607 (2014-08-19)
Theoretical study on the structural and vibrational analysis of monomer and dimer structures of 2,4,5-trimethylbenzoic acid (2,4,5-TMBA, C₁₀H₁₂O₂) were presented. The geometry of the molecule was fully optimized. The Fourier transform infrared (FT-IR) and the Fourier transform Raman (FT-Raman) spectra
2, 4, 5-Trimethylbenzoic Acid.
Barcon A, et al.
Acta Crystallographica Section C, Structural Chemistry, 53(12), 1842-1845 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service