Skip to Content
MilliporeSigma
All Photos(1)

Documents

526088

Sigma-Aldrich

Diethyl 4-methoxybenzylphosphonate

≥98%

Synonym(s):

[(4-Methoxyphenyl)methyl]phosphonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
258.25
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

Assay

≥98%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.504 (lit.)

bp

155 °C/0.5 mmHg (lit.)

density

1.132 g/mL at 25 °C (lit.)

SMILES string

CCOP(=O)(Cc1ccc(OC)cc1)OCC

InChI

1S/C12H19O4P/c1-4-15-17(13,16-5-2)10-11-6-8-12(14-3)9-7-11/h6-9H,4-5,10H2,1-3H3

InChI key

NKARVHNAACNYGE-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • γ -Monofluorinated goniothalamin analogs via ring-opening hydrofluorination
  • Stilbenoid derivatives with neuroprotective activity
  • Resveratrol-chroman hybrids with antioxidant activity
  • Oligostilbenoids using regioselective cyclodehydration and arylation
  • Resveratrol fatty alcohols with effects on differentiation of neural stem cells and modulation of neuroinflammation

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service