Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

526053

Sigma-Aldrich

Diethyl-4-methylbenzylphosphonate

97%

Synonym(s):

Diethyl p-tolylmethylphosphonate, [(4-Methylphenyl)methyl]phosphonic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
242.25
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

Assay

97%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.497 (lit.)

bp

110 °C/0.2 mmHg (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

CCOP(=O)(Cc1ccc(C)cc1)OCC

InChI

1S/C12H19O3P/c1-4-14-16(13,15-5-2)10-12-8-6-11(3)7-9-12/h6-9H,4-5,10H2,1-3H3

InChI key

QKGBKPZAXXBLJE-UHFFFAOYSA-N

Application

Reactant involved in:
  • Synthesis of stilbenes via oxidation and Horner-Emmons reactions
  • Intramolecular cyclization of aryl ethers, amines, and amides
  • Studies of the effects of functional groups on the performance of blue organic LEDs
  • Olefination reactions
  • Preparation of antimalarial agents

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service