Skip to Content
MilliporeSigma
All Photos(1)

Documents

525588

Sigma-Aldrich

5-Amino-2-methylindole

97%

Synonym(s):

2-Methylindol-5-ylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10N2
CAS Number:
Molecular Weight:
146.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

153-157 °C (lit.)

SMILES string

Cc1cc2cc(N)ccc2[nH]1

InChI

1S/C9H10N2/c1-6-4-7-5-8(10)2-3-9(7)11-6/h2-5,11H,10H2,1H3

InChI key

JQULCCZIXYRBSE-UHFFFAOYSA-N

Application

Reactant for preparation of:
  • Cytotoxic and antimitotic agents
  • Potential antimicrobial aminoketones
  • Inhibitors of monoamine oxidase (MAO)
  • Protein kinase c theta (PKCθ) inhibitors
  • Factor Xa inhibitors
  • Inhibitors of vascular endothelial growth factor type 2 (VEGFR-2)
  • Demethylasterriquinone B1receptor ligands

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niina Tani et al.
Bioorganic & medicinal chemistry, 22(23), 6655-6664 (2014-12-03)
Inhibition of CYP2A6-mediated nicotine metabolism can reduce cigarette smoking. We sought potent and selective CYP2A6 inhibitors to be used as leads for drugs useful in smoking reduction therapy, by evaluating CYP2A6 inhibitory effect of novel formyl, alkyl amine or carbonitrile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service