Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

525154

Sigma-Aldrich

Dioctyl phthalate

99%

Synonym(s):

DOP, Bis(2-ethylhexyl) phthalate, DEHP, Phthalic acid bis(2-ethylhexyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-[CO2CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
390.56
Beilstein:
1890696
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

>16 (vs air)

vapor pressure

1.2 mmHg ( 93 °C)

Assay

99%

autoignition temp.

734 °F

refractive index

n20/D 1.486 (lit.)

bp

384 °C (lit.)

mp

−50 °C (lit.)

density

0.985 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)COC(=O)c1ccccc1C(=O)OCC(CC)CCCC

InChI

1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3

InChI key

BJQHLKABXJIVAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Plasticizer

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

404.6 °F - closed cup

Flash Point(C)

207 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mohammad Mahdi Najafpour et al.
Dalton transactions (Cambridge, England : 2003), 43(34), 13122-13135 (2014-07-22)
Eighteen Mn complexes with N-donor and carboxylate ligands have been synthesized and characterized. Three Mn complexes among them are new and are reported for the first time. The reactions of oxygen evolution in the presence of oxone (2KHSO5·KHSO4·K2SO4) and cerium(iv)
Minjin Lee et al.
International journal of environmental research and public health, 15(4) (2018-04-14)
The inhalation of a water aerosol from a humidifier containing disinfectants has led to serious lung injuries in Korea. To promote the safe use of products, the Korean government enacted regulations on the chemicals in various consumer products that could
Anne-Marie Saillenfait et al.
Journal of applied toxicology : JAT, 33(9), 1027-1035 (2013-06-12)
In utero exposure to the phthalate ester plasticizer di-n-hexyl phthalate (DnHP) is known to affect the development of the male reproductive system and induce alterations in androgen-dependent tissues of male rat offspring. Male reproductive malformations produced by several phthalates have
N B Hopf et al.
Toxicology letters, 224(1), 47-53 (2013-10-22)
Phthalates are suspected to be endocrine disruptors. Di(2-ethylhexyl) phthalate (DEHP) is assumed to have low dermal absorption; however, previous in vitro skin permeation studies have shown large permeation differences. Our aims were to determine DEHP permeation parameters and assess extent
Na Rae Song et al.
Environment international, 54, 65-73 (2013-02-19)
Di(2-ethylhexyl) phthalate (DEHP) is one of the common phthalate plasticizers used primarily in soft polyvinyl chloride, which is a plastic polymer that makes up the total weight of goods from 1% up to 40% in many consumer products. The aims

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service