Skip to Content
MilliporeSigma
All Photos(2)

Documents

524883

Sigma-Aldrich

1-(Dimethylsulfamoyl)imidazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9N3O2S
CAS Number:
Molecular Weight:
175.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

48-51 °C (lit.)

SMILES string

CN(C)S(=O)(=O)n1ccnc1

InChI

1S/C5H9N3O2S/c1-7(2)11(9,10)8-4-3-6-5-8/h3-5H,1-2H3

InChI key

YRRWNBMOJMMXQY-UHFFFAOYSA-N

General description

1-(Dimethylsulfamoyl)imidazole is an imidazole derivative.

Application

1-(Dimethylsulfamoyl)imidazole may be used to synthesize 1-(dimethylsulfamoyl)-2-imidazolecarboxaldehyde.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 1-(dimethylsulfamoyl)-2-and 5-imidazolecarboxaldehydes. Rearrangement of 1-(dimethylsulfamoyl)-5-imidazole-carboxaldehyde to the 4-carboxaldehyde.
Kim JW, et al.
Journal of Heterocyclic Chemistry, 32(2), 611- 620 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service