Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

524700

Sigma-Aldrich

3,4,5-Trifluorophenylboronic acid

≥95%

Synonym(s):

3,4,5-Trifluorobenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3C6H2B(OH)2
CAS Number:
Molecular Weight:
175.90
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 524700, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

≥95%

form

powder

mp

290-295 °C (lit.)

functional group

fluoro

SMILES string

OB(O)c1cc(F)c(F)c(F)c1

InChI

1S/C6H4BF3O2/c8-4-1-3(7(11)12)2-5(9)6(4)10/h1-2,11-12H

InChI key

UHDDEIOYXFXNNJ-UHFFFAOYSA-N

Application

Reactant involved in:
  • Preparation of phenylboronic catechol esters and determination of Lewis acidity
  • Synthesis of benzopyranone derivatives as GABAA receptor modulators
  • Synthesis of multisubstituted olefins and conjugate dienes
  • Suzuki cross-coupling reactions
  • Preparation of fluorinated aromatic poly(ether-amide)s

Other Notes

Contains varying amounts of anhydride.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mikhail Y Vorona et al.
Materials (Basel, Switzerland), 13(8) (2020-04-26)
Anthracene-based semiconductors have attracted great interest due to their molecular planarity, ambient and thermal stability, tunable frontier molecular orbitals and strong intermolecular interactions that can lead to good device field-effect transistor performance. In this study, we report the synthesis of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service