Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

524697

Sigma-Aldrich

2,3,5-Trifluorophenylboronic acid

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3C6H2B(OH)2
CAS Number:
Molecular Weight:
175.90
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

solid

mp

221 °C (dec.) (lit.)

functional group

fluoro

SMILES string

OB(O)c1cc(F)cc(F)c1F

InChI

1S/C6H4BF3O2/c8-3-1-4(7(11)12)6(10)5(9)2-3/h1-2,11-12H

InChI key

IRMUMGKQAXLGHK-UHFFFAOYSA-N

Application

Reactant involved in the synthesis of biologically active compounds:
  • Pyrazole-acids to be used as agonists of human orphan GPCR GPR109a
  • Bicyclic pyrazole carboxylic acid derivatives as a niacin receptor agonist
  • 5-Alkyl and 5-aryl-pyrazole-tetrazoles as agonists of human orphan GPCR GPR109a

Other Notes

Contains varying amounts of anhydride

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service