Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

523666

Sigma-Aldrich

3-(Trimethylsilyl)phenylboronic acid

≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiC6H4B(OH)2
CAS Number:
Molecular Weight:
194.11
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

form

solid

mp

145-150 °C (lit.)

SMILES string

C[Si](C)(C)c1cccc(c1)B(O)O

InChI

1S/C9H15BO2Si/c1-13(2,3)9-6-4-5-8(7-9)10(11)12/h4-7,11-12H,1-3H3

InChI key

REMKRZLFPLDTKR-UHFFFAOYSA-N

Application

3-(Trimethylsilyl)phenylboronic acid can be used:
  • To prepare a 2-phenylpyridine ligand, which is used in the synthesis of Ir(III) complex applicable in the OLEDs.
  • As an intermediate in the preparation of biphenyl derived amino acids.
  • As a starting material for the synthesis of triphenylene-based electron transport materials.

Other Notes

Contains varying amounts of anhydride

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient and practical synthesis of electron transport material and its key intermediate
Zhao X, et al.
Organic Process Research & Development, 21(10), 1675-1681 (2017)
Synthesis and hydrogen bonding capabilities of biphenyl-based amino acids designed to nucleate β-sheet structure
Nesloney CL and Kelly JW
The Journal of Organic Chemistry, 61(9), 3127-3137 (1996)
A green emitting iridium (III) complex with narrow emission band and its application to phosphorescence organic light-emitting diodes (OLEDs)
Jung SO, et al.
Organic Electronics, 10(6), 1066-1073 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service