Skip to Content
MilliporeSigma
All Photos(1)

Documents

523208

Sigma-Aldrich

Methyl-d3-triphenylphosphonium iodide

95 atom % D

Synonym(s):

Triphenyl(trideuteriomethyl)phosphonium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3P(C6H5)3I
CAS Number:
Molecular Weight:
407.24
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

95 atom % D

reaction suitability

reaction type: C-C Bond Formation
reaction type: Whiting Reaction

mp

186-188 °C (lit.)

mass shift

M+3

SMILES string

[I-].[2H]C([2H])([2H])[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C19H18P.HI/c1-20(17-11-5-2-6-12-17,18-13-7-3-8-14-18)19-15-9-4-10-16-19;/h2-16H,1H3;1H/q+1;/p-1/i1D3;

InChI key

JNMIXMFEVJHFNY-NIIDSAIPSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service