Skip to Content
MilliporeSigma
All Photos(1)

Documents

522546

Sigma-Aldrich

1,8-Dihydroxy-4,5-dinitroanthraquinone

97%

Synonym(s):

ARDP0006

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H6N2O8
CAS Number:
Molecular Weight:
330.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

225 °C (dec.) (lit.)

SMILES string

Oc1ccc(c2C(=O)c3c(ccc(O)c3C(=O)c12)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C14H6N2O8/c17-7-3-1-5(15(21)22)9-11(7)14(20)12-8(18)4-2-6(16(23)24)10(12)13(9)19/h1-4,17-18H

InChI key

GJCHQJDEYFYWER-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement and Correlation of Derivatized Anthraquinone Solubility in Supercritical Carbon Dioxide.
Alwi RS and Tamura K.
Journal of Chemical and Engineering Data, 60(10), 3046-3052 (2015)
Catalysed hydrogenation of hydroxynitroanthraquinones in strong acid.
Morley JO and Hutcheson R.
Journal of Chemical Technology and Biotechnology, 30(1), 409-414 (1980)
David A Constant et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(40), 10136-10141 (2018-09-20)
Many positive-strand RNA viruses translate their genomes as single polyproteins that are processed by host and viral proteinases to generate all viral protein products. Among these is dengue virus, which encodes the serine proteinase NS2B/3 responsible for seven different cleavages

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service