Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

521639

Sigma-Aldrich

6,11-Dihydrodibenzo[b,e]thiepin-11-one

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10OS
CAS Number:
Molecular Weight:
226.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

86-88 °C (lit.)

SMILES string

O=C1c2ccccc2CSc3ccccc13

InChI

1S/C14H10OS/c15-14-11-6-2-1-5-10(11)9-16-13-8-4-3-7-12(13)14/h1-8H,9H2

InChI key

JGJDEWXZEIHBNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6,11-Dihydrodibenzo[b,e]thiepin-11-one is an organic building block. Its density is 1.2151g/ml at 25°C.[1]

Application

6,11-Dihydrodibenzo[b,e]thiepin-11-one may be used as a starting reagent in the synthesis of 6,11-dihydrodibenzo[b,e]thiepin-11-one 5,5-dioxide.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 325-325 (2014)
Synthesis, characterization and antimicrobial activity evaluation of some new derivatives of 6, 11-dihydrodibenzo [b,e] thiepin 5, 5-dioxide.
Stecoza CE, et al.
Revista de Chimie (Bucharest, Romania), 60(2), 137-141 (2009)
Zdena Křesinová et al.
Journal of chromatography. A, 1440, 15-22 (2016-03-01)
A rapid and reliable analytical method was developed for the quantitative determination of psychopharmaceuticals, their precursors and by-products in real contaminated samples from a pharmaceutical company in Olomouc (Czech Republic), based on SPE disk extraction and detection by ultra performance

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service