Skip to Content
MilliporeSigma
All Photos(2)

Documents

520845

Sigma-Aldrich

Palladium on carbon

greener alternative

extent of labeling: 5 wt. % loading, matrix carbon powder, wet support

Synonym(s):

Escat 11, Pd/C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
Molecular Weight:
106.42
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

5 wt. % loading

matrix

carbon powder, wet support

greener alternative category

SMILES string

[Pd]

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Catalyst for:
  • Stille reaction
  • Hydrogenation reactions
  • Suzuki-Miyaura cross-coupling and Heck-Mizoroki reactions
  • Deoxygenation
  • Oxidation reactions
  • Coupling reactions
Palladium on carbon (Pd/C) (5 wt. % loading, matrix carbon powder, wet support) can be used for a variety of applications, such as:
  • a catalyst for hydrodechlorination of dichlorodifluoromethane (CCl2F2), which is a known greenhouse gas
  • a catalyst for Heck reactions of aryl halides in ionic liquids
  • an anode catalyst for the fabrication of direct formic acid fuel cells (DFAFC)
  • a cost-effective electrocatalyst for the oxidation of methanol
  • in the catalytic hydrogenation of chemical hydrides for the production of hydrogen
  • a catalyst in Suzuki-Mayaura reaction
Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Legal Information

ESCAT is a trademark of BASF Catalysts, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heterogeneous Heck reaction catalyzed by Pd/C in ionic liquid
Hagiwara H, et al.
Tetrahedron Letters, 42(26), 4349-4351 (2001)
Pd/C: an old catalyst for new applications-its use for the Suzuki-Miyaura reaction
Felpin F, et al.
European Journal of Organic Chemistry, 2006(12), 2679-2690 (2006)
Pd-C powder and thin film catalysts for hydrogen production by hydrolysis of sodium borohydride
Patel N, et al.
International Journal of Hydrogen Energy, 33(1), 287-292 (2008)
Characterisation of the deactivation of platinum and palladium supported on activated carbon used as hydrodechlorination catalysts
Ordonez S, et al.
Applied Catalysis. B, Environmental, 31(2), 113-122 (2001)
Performance characterization of Pd/C nanocatalyst for direct formic acid fuel cells
Ha S, et al.
Journal of Power Sources, 144(1), 28-34 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service