Skip to Content
MilliporeSigma
All Photos(2)

Documents

520586

Sigma-Aldrich

Dibutyltin bis(acetylacetonate)

95%

Synonym(s):

Sn(acac)Bu2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3COCH=C(O-)CH3]2Sn[(CH2)3CH3]2
CAS Number:
Molecular Weight:
431.15
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

low-melting solid

reaction suitability

core: tin

refractive index

n20/D 1.5110 (lit.)

bp

150 °C (lit.)

mp

26 °C (lit.)

density

1.22 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.2C4H9.Sn/c2*1-4(6)3-5(2)7;2*1-3-4-2;/h2*3,6H,1-2H3;2*1,3-4H2,2H3;/q;;;;+2/p-2/b2*4-3-;;;

InChI key

UVDDHYAAWVNATK-VGKOASNMSA-L

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Sol. 2 - Muta. 2 - Repr. 1B - Skin Corr. 1C - Skin Sens. 1 - STOT RE 1 Oral - STOT SE 1

Target Organs

thymus gland

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

130.0 °F - closed cup

Flash Point(C)

54.44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service