Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

520578

Sigma-Aldrich

2-Phenylthiophene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8S
CAS Number:
Molecular Weight:
160.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

bp

256 °C (lit.)

mp

34-36 °C (lit.)

functional group

phenyl

storage temp.

2-8°C

SMILES string

c1ccc(cc1)-c2cccs2

InChI

1S/C10H8S/c1-2-5-9(6-3-1)10-7-4-8-11-10/h1-8H

InChI key

PJRGDKFLFAYRBV-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Some Substitution Reactions of 2-Phenylthiophene.
Gjos N and Gronowit S.
Acta Chemica Scandinavica, 26, 1851-1859 (1972)
Microwave-assisted synthesis of 2-acetyl-5-arylthiophenes and 4-(5-arylthiophen-2-yl) thiazoles via Suzuki coupling in water.
Dawood KM, et al.
ARKIVOC (Gainesville, FL, United States), 7, 50-62 (2015)
On the nitration of 2-and 3-phenylthiophene.
Gronowitz SALO and Gios N.
Acta Chemica Scandinavica, 21(10), 16-16 (1967)
Ji He et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 16(7), 1049-1056 (2017-05-13)
Three D-D-π-A triphenylamine-coumarin sensitizers were designed and synthesized with a triphenylamine and coumarin double donor and a cyanoacrylic acid acceptor. Thiophene, bithiophene and phenylthiophene were used as π-bridges, respectively, and the effects of different π-bridges on the photophysical properties and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service