Skip to Content
MilliporeSigma
All Photos(2)

Documents

520438

Sigma-Aldrich

5-Bromo-2-fluoropyridine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3BrFN
CAS Number:
Molecular Weight:
175.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.5325 (lit.)

bp

162-164 °C/750 mmHg (lit.)

density

1.71 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(Br)cn1

InChI

1S/C5H3BrFN/c6-4-1-2-5(7)8-3-4/h1-3H

InChI key

MYUQKYGWKHTRPG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew Sutherland et al.
The Journal of organic chemistry, 68(6), 2475-2478 (2003-03-15)
Two racemic fluoropyridine analogues 4 and 5 of the potent nicotinic agonist UB-165 have been synthesized. Halogenated pyridines 7 and 12 provided the organometallic reagents needed for the Negishi and Suzuki coupling reactions used for the preparation of 4 and
Andrew Sutherland et al.
The Journal of organic chemistry, 68(8), 3352-3355 (2003-04-12)
5-bromo-2-fluoro-3-pyridylboronic acid (3) was prepared in high yield by ortho-lithiation of 5-bromo-2-fluoropyridine (1), followed by reaction with trimethylborate. Suzuki reaction of 3 with a range of aryl iodides gave 3-monosubstituted 5-bromo-2-fluoropyridines 4 in excellent yields. A second Suzuki reaction utilizing
Aromatic fluorine compounds. XI. Replacement of chlorine by fluorine in halopyridines.
Finger GC, et al.
The Journal of Organic Chemistry, 28(6), 1666-1668 (1963)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service