Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

519367

Sigma-Aldrich

3,5-Bis(trifluoromethyl)benzenesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2C6H3SO2Cl
CAS Number:
Molecular Weight:
312.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

34-38 °C (lit.)

SMILES string

FC(F)(F)c1cc(cc(c1)S(Cl)(=O)=O)C(F)(F)F

InChI

1S/C8H3ClF6O2S/c9-18(16,17)6-2-4(7(10,11)12)1-5(3-6)8(13,14)15/h1-3H

InChI key

BTRCVKADYDVSLI-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly enantioselective catalytic thiolysis of prochiral cyclic dicarboxylic anhydrides utilizing a bifunctional chiral sulfonamide.
Takashi Honjo et al.
Angewandte Chemie (International ed. in English), 44(36), 5838-5841 (2005-08-05)
Etienne Borré et al.
Beilstein journal of organic chemistry, 6, 1159-1166 (2010-12-18)
Seven novel Hoveyda-Grubbs precatalysts bearing an aminosulfonyl function are reported. Kinetic studies indicate an activity enhancement compared to Hoveyda's precatalyst. A selection of these catalysts was investigated with various substrates in ring-closing metathesis of dienes or enynes and cross metathesis.
Protein diffusion in porous gel filtration chromatography media studied by pulsed field gradient NMR spectroscopy.
Gibbs SJ, et al.
The Journal of Physical Chemistry, 96(18), 7458-7462 (1992)
Versatile chiral reagent for the highly enantioselective synthesis of either anti or syn ester aldols.
Corey EJ and Kim SS.
Journal of the American Chemical Society, 112(12), 4976-4977 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service