Skip to Content
MilliporeSigma
All Photos(1)

Documents

519359

Sigma-Aldrich

2-Fluoro-6-iodobenzonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(I)CN
CAS Number:
Molecular Weight:
247.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

49-53 °C (lit.)

SMILES string

Fc1cccc(I)c1C#N

InChI

1S/C7H3FIN/c8-6-2-1-3-7(9)5(6)4-10/h1-3H

InChI key

FAACTMVXBNSPJA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent advances in the chemistry of indazoles.
Schmidt A, et al.
European Journal of Organic Chemistry, 2008(24), 4073-4095 (2008)
2-Fluoro-6-iodobenzonitrile.
Britton D.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 57(8), o702-o704 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service