Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

518948

Sigma-Aldrich

2-Chloro-3-methylpyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6ClN
CAS Number:
Molecular Weight:
127.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.533 (lit.)

bp

192-193 °C/751 mmHg (lit.)

density

1.17 g/mL at 25 °C (lit.)

SMILES string

Cc1cccnc1Cl

InChI

1S/C6H6ClN/c1-5-3-2-4-8-6(5)7/h2-4H,1H3

InChI key

RKVUCIFREKHYTL-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

175.0 °F - closed cup

Flash Point(C)

79.44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study on Synthetic Technology of 2-Chloro-3-Pyridinecarboxlic acid [J].
Wen-na NIE.
Hebei Chemical Industry, 10, 004-004 (2006)
Thomas J Colacot et al.
Organic letters, 6(21), 3731-3734 (2004-10-08)
[reaction: see text] The use of Cp(2)Fe(PR(2))(2)PdCl(2) (R = i-Pr and t-Bu) in Suzuki coupling reactions were illustrated using a high throughput screening approach. The di-tbpfPdCl(2) catalyst was shown to be the more active catalyst for unactivated and sterically challenging

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service