Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

518875

Sigma-Aldrich

2-Nitrocyclododecanone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC12H21(=O)
CAS Number:
Molecular Weight:
227.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

77-80 °C (lit.)

SMILES string

[O-][N+](=O)C1CCCCCCCCCCC1=O

InChI

1S/C12H21NO3/c14-12-10-8-6-4-2-1-3-5-7-9-11(12)13(15)16/h11H,1-10H2

InChI key

SIKBTEDZSLYKRA-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Juan Zhu et al.
Journal of agricultural and food chemistry, 56(15), 6547-6553 (2008-07-12)
Four series of novel macrolactones and macrolactams12-alkylsulfonamido-1,15-pentadecanlactones ( 5), 12-alkylsulfonamido-15-methyl-1,15-pentadecanlactones ( 6), 12-alkylsulfonamido-1,15-pentadecanlactams ( 7), and N-(alkylsulfonamidoethyl)-1,12-dodecanlactams ( 8)were designed and synthesized from readily available 2-nitrocyclododecanone or cyclododecanone. Their structures were confirmed by (1)H NMR, IR, and elemental analysis. The
The Alkyl Nitration of Active Methylene Compounds. IV. The Mononitration of Ketones.
Feuer H and Pivawer PM.
The Journal of Organic Chemistry, 31(10), 3152-3158 (1966)
Synthesis of 15-Cyano-12-oxopentadecano-15-lactone and 15-Cyano-12-oxopentadecano-15-lactam.
Wu P, et al.
Helvetica Chimica Acta, 92(12), 2774-2782 (2009)
A short synthesis of 15-pentadecanolide.
Stanchev S, et al.
Tetrahedron Letters, 34(38), 6107-6108 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service