Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

518360

Sigma-Aldrich

Boc-Hyp-OH

97%

Synonym(s):

trans-N-(tert-Butoxycarbonyl)-4-hydroxy-L-proline, Boc-L-hydroxyproline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO5
CAS Number:
Molecular Weight:
231.25
Beilstein:
4295484
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D −78° in H2O

mp

123-127 °C (lit.)

SMILES string

CC(C)(C)OC(=O)N1C[C@H](O)C[C@H]1C(O)=O

InChI

1S/C10H17NO5/c1-10(2,3)16-9(15)11-5-6(12)4-7(11)8(13)14/h6-7,12H,4-5H2,1-3H3,(H,13,14)/t6-,7+/m1/s1

InChI key

BENKAPCDIOILGV-RQJHMYQMSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service