Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

518328

Sigma-Aldrich

4-Methyl-3-nitrobenzaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)CHO
CAS Number:
Molecular Weight:
165.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

46-49 °C (lit.)

functional group

aldehyde
nitro

SMILES string

Cc1ccc(C=O)cc1[N+]([O-])=O

InChI

1S/C8H7NO3/c1-6-2-3-7(5-10)4-8(6)9(11)12/h2-5H,1H3

InChI key

KHWGAWBXQOKXIJ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The chemistry of indoles. XXXII A facile synthetic method for 6-indolecarbaldehyde, 6-indolemethanol, and 6-substituted 1-hydroxyindoles and its application for the synthesis of a natural alkaloid,(E)-6-(3-methylbuta-1, 3-dienyl) indole.
Somei M.
Chemical & Pharmaceutical Bulletin, 34(10), 4109-4115 (1986)
Study of vibrational spectra of 4-methyl-3-nitrobenzaldehyde.
Yadav BS, et al.
Indian Journal of Pure and Applied Physics, 44(9), 644-644 (2006)
Total synthesis of mimosamycin.
Fukumi H, et al.
Chemical & Pharmaceutical Bulletin, 26(7), 2175-2180 (1978)
4-Methyl-3-nitrobenzaldehyde.
Guo ZR, et al.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 66(9), o2420-o2420 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service