Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

518115

Sigma-Aldrich

3-(2-Aminoethyl)pyridine dihydrobromide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2 · 2HBr
CAS Number:
Molecular Weight:
283.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

248-252 °C (lit.)

functional group

amine

SMILES string

Br.Br.NCCc1cccnc1

InChI

1S/C7H10N2.2BrH/c8-4-3-7-2-1-5-9-6-7;;/h1-2,5-6H,3-4,8H2;2*1H

InChI key

FQKYVSOKWFXVFX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Willem M Albers et al.
International journal of molecular sciences, 14(2), 3500-3513 (2013-02-08)
Pyridyl derivatives of lipoic acid were prepared as ligands for the study of the interaction with thyroxine (T4). Thin self-assembled films of the ligands were prepared in 70% ethanol on gold and their interaction with T4 was studied by titration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service