Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

518042

Sigma-Aldrich

4-Butylbenzaldehyde

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3C6H4CHO
CAS Number:
Molecular Weight:
162.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

refractive index

n20/D 1.522 (lit.)

bp

250-263 °C (lit.)

density

0.968 g/mL at 25 °C (lit.)

SMILES string

CCCCc1ccc(C=O)cc1

InChI

1S/C11H14O/c1-2-3-4-10-5-7-11(9-12)8-6-10/h5-9H,2-4H2,1H3

InChI key

ARIREUPIXAKDAY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Central linkage influence upon mesomorphic and electrooptical behavior of diaryl nematics. General proton magnetic resonance method employing a lanthanide shift reagent for analysis of isomeric azoxybenzenes.
Rondeau RE, et al.
Journal of the American Chemical Society, 94(4), 1096-1102 (1972)
Francis Giraud et al.
Journal of enzyme inhibition and medicinal chemistry, 24(5), 1067-1075 (2009-06-27)
A new series of 1-benzyl-3-(imidazol-1-ylmethyl)indoles were synthesized according to a previous 3D-QSAR predictive model and assayed for their antiparasitic activity upon Leishmania mexicana promastigotes. The biological results obtained for these twelve molecules showed an IC(50) ranging from 2.3 to 32
Chao-Bin Xue et al.
Bioorganic & medicinal chemistry, 15(5), 2006-2015 (2007-01-30)
Phenoloxidase (PO), also known as tyrosinase, is a key enzyme in insect development, responsible for catalyzing the hydroxylation of tyrosine into o-diphenols and the oxidation of o-diphenols into o-quinones. Inhibition of PO may provide a basis for novel environmentally friendly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service