Skip to Content
MilliporeSigma
All Photos(1)

Documents

517941

Sigma-Aldrich

Poly(acrylonitrile-co-methyl acrylate)

acrylonitrile ~94 wt. %

Synonym(s):

Acrylic acid methyl ester, polymer with acrylonitrile, Acrylonitrile-methyl acrylate copolymer, Acrylonitrile-methyl acrylate polymer, Methyl acrylate-acrylonitrile copolymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CN)]x[CH2CH(CO2CH3)]y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

composition

acrylonitrile, ~94 wt. %

SMILES string

C=CC#N.COC(=O)C=C

Related Categories

Application

Poly(acrylonitrile-co-methyl acrylate) (P(AN-co-MA)) may be used to fabricate multilayer mixed matrix membranes (MMMMs) for easy separation of ethanol from an ethanol-water mixture.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly (vinyl alcohol) multilayer mixed matrix membranes for the dehydration of ethanol?water mixture.
Guan H M, et al.
Journal of Membrane Science , 268(2), 113-122 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service