Skip to Content
MilliporeSigma
All Photos(1)

Documents

516880

Sigma-Aldrich

(2,6-Dimethyl-4H-pyran-4-ylidene)malononitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2O
CAS Number:
Molecular Weight:
172.18
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

99%

mp

192-194 °C (lit.)

SMILES string

CC1=C\C(C=C(C)O1)=C(\C#N)C#N

Application

Dye intermediate used in the synthesis of an electrooptical chromophore.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beckmann, S. et al.
Advanced Materials, 11, 536-536 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service