Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

516716

Sigma-Aldrich

Ethanolamine dithiodipropionate solution

75 wt. % in H2O

Synonym(s):

TPS® 15

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H24N2O6S2
CAS Number:
Molecular Weight:
332.44
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

concentration

75 wt. % in H2O

SMILES string

NCCO.NCCO.OC(=O)CCSSCCC(O)=O

InChI

1S/C6H10O4S2.2C2H7NO/c7-5(8)1-3-11-12-4-2-6(9)10;2*3-1-2-4/h1-4H2,(H,7,8)(H,9,10);2*4H,1-3H2

InChI key

OBJGBTROGRUPPA-UHFFFAOYSA-N

Legal Information

Product of Arkema
TPS is a registered trademark of Arkema France

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service