Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

516295

Sigma-Aldrich

1-Pentadecanethiol

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14SH
CAS Number:
Molecular Weight:
244.48
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

≥97%

refractive index

n20/D 1.462 (lit.)

bp

325 °C (lit.)

density

0.847 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCS

InChI

1S/C15H32S/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16/h16H,2-15H2,1H3

InChI key

IGMQODZGDORXEN-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hongseung Yeon et al.
Langmuir : the ACS journal of surfaces and colloids, 33(19), 4628-4637 (2017-04-20)
We report an experimental investigation of the influence of molecular-level order (crystallinity) within nonpolar monolayers on hydrophobic interactions. The measurements were performed using gold film-supported monolayers formed from alkanethiols (CH
Alif Chebbi et al.
FEMS microbiology letters, 362(14) (2015-06-19)
Hydrogen sulfide (H2S) and thiols (RSH) generated by the phosphate industry cause harmful effects on human health and quality of life. The present study aims to investigate and evaluate a bacterial strain CAT37 isolated from gas-washing wastewaters in terms of

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service