Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

516147

Sigma-Aldrich

6-Methyl-2,2′-dipyridyl

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10N2
CAS Number:
Molecular Weight:
170.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.607 (lit.)

bp

117-119 °C/0.1 mmHg (lit.)

density

1.09 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(n1)-c2ccccn2

InChI

1S/C11H10N2/c1-9-5-4-7-11(13-9)10-6-2-3-8-12-10/h2-8H,1H3

InChI key

LLCYXFYLGPOKQO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient synthesis of 4-, 5-, and 6-methyl-2, 2'-bipyridine by a Negishi cross-coupling strategy followed by high-yield conversion to bromo-and chloromethyl-2, 2'-bipyridines.
Savage SA, et al.
The Journal of Organic Chemistry, 63(26), 10048-10051 (1998)
Bobo Skillinghaug et al.
The Journal of organic chemistry, 79(24), 12018-12032 (2014-10-09)
A fast and efficient protocol for the palladium(II)-catalyzed production of aryl ketones from sodium arylsulfinates and various organic nitriles under controlled microwave irradiation has been developed. The wide scope of the reaction has been demonstrated by combining 14 sodium arylsulfinates
Chemical Constitution and Activity of Bipyridylium Herbicides. XII. Diquaternary Salts of 6-Methyl-2, 2'-bipyridyl and 2-Methyl-4, 4'-bipyridyl.
Schmalzl K and Summers L.
Australian Journal of Chemistry, 30(3), 657-662 (1977)
Synthesis, spectroscopy, and electrochemistry of ruthenium (II) complexes of 6-(N-pyrazolyl)-and 6-(N-pyrazolylmethyl)-2, 2'-bipyridines: new tridentate ligands.
Downard AJ, et al.
Inorganic Chemistry, 30(91), 3733-3737 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service