Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

515930

Sigma-Aldrich

Nitrosonium hexafluoroantimonate

99.9% trace metals basis

Synonym(s):

Nitrosonium hexafluoroantimonate, Nitrosyl hexafluoroantimonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NOSbF6
CAS Number:
Molecular Weight:
265.76
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

99.9% trace metals basis

form

powder

impurities

≤1500.0 ppm Trace Metal Analysis

SMILES string

N#[O+].F[Sb-](F)(F)(F)(F)F

InChI

1S/6FH.NO.Sb/c;;;;;;1-2;/h6*1H;;/q;;;;;;+1;+5/p-6

InChI key

AMGJCGWYCZRXHS-UHFFFAOYSA-H

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service