Skip to Content
MilliporeSigma
All Photos(1)

Documents

515817

Sigma-Aldrich

Zirconium(IV) bis(diethyl citrato)dipropoxide

Synonym(s):

TYZOR® ZEC organic zirconate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44O16Zr
CAS Number:
Molecular Weight:
703.84
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

liquid

reaction suitability

core: zirconium

impurities

12% ethanol and 1-propanol

refractive index

n20/D 1.451 (lit.)

density

1.14 g/mL at 25 °C (lit.)

SMILES string

CCCO[Zr](OCCC)(OC(=O)C(O)(CC(=O)OCC)CC(=O)OCC)OC(=O)C(O)(CC(=O)OCC)CC(=O)OCC

InChI

1S/2C10H16O7.2C3H7O.Zr/c2*1-3-16-7(11)5-10(15,9(13)14)6-8(12)17-4-2;2*1-2-3-4;/h2*15H,3-6H2,1-2H3,(H,13,14);2*2-3H2,1H3;/q;;2*-1;+4/p-2

InChI key

JNNLYJHFWHGPFC-UHFFFAOYSA-L

Legal Information

TYZOR is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

65.0 °F

Flash Point(C)

18.33 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service