Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

515108

Sigma-Aldrich

5-Methoxybenzimidazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N2O
CAS Number:
Molecular Weight:
148.16
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
32151902
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

121-126 °C (lit.)

SMILES string

COc1ccc2[nH]cnc2c1

InChI

1S/C8H8N2O/c1-11-6-2-3-7-8(4-6)10-5-9-7/h2-5H,1H3,(H,9,10)

InChI key

ILMHAGCURJPNRZ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristi Kincaid et al.
Nucleic acids research, 33(8), 2620-2628 (2005-05-10)
In order to further understand how DNA polymerases discriminate against incorrect dNTPs, we synthesized two sets of dNTP analogues and tested them as substrates for DNA polymerase alpha (pol alpha) and Klenow fragment (exo-) of DNA polymerase I (Escherichia coli).
On the Biosynthesis of 5-Methoxybenzimidazole.
Wurm R, et al.
European Journal of Biochemistry, 56(2), 427-432 (1975)
Computational determination of aqueous p K a values of protonated benzimidazoles (Part 2).
Brown TN and Mora-Diez N.
The Journal of Physical Chemistry B, 110(41), 20546-20554 (2006)
Structural investigation of the biosynthesis of alternative lower ligands for cobamides by nicotinate mononucleotide: 5, 6-dimethylbenzimidazole phosphoribosyltransferase from Salmonella enterica.
Cheong C, et al.
The Journal of Biological Chemistry, 276(40), 37612-37620 (2001)
An exceptionally mild catalytic thioester aldol reaction inspired by polyketide biosynthesis.
Lalic G, et al.
Journal of the American Chemical Society, 125(10), 2852-2853 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service