Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

514047

Sigma-Aldrich

2,3-Dichlorophenylboronic acid

Synonym(s):

(2,3-Dichlorophenyl)dihydroxyborane, 2,3-Dichlorobenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3B(OH)2
CAS Number:
Molecular Weight:
190.82
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

mp

125 °C (dec.) (lit.)

functional group

chloro

SMILES string

OB(O)c1cccc(Cl)c1Cl

InChI

1S/C6H5BCl2O2/c8-5-3-1-2-4(6(5)9)7(10)11/h1-3,10-11H

InChI key

TYIKXPOMOYDGCS-UHFFFAOYSA-N

Application

Reactant for:
  • Suzuki-Miyaura coupling reactions
  • Preparation of biologically and pharmacologically active molecules

Other Notes

Contains varying amounts of anhydride

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xueshu Li et al.
Environmental science and pollution research international, 25(17), 16402-16410 (2017-05-26)
Nineteen polychlorinated biphenyl (PCB) congeners, such as 2,2',3,3',6-pentachlorobiphenyl (PCB 84), display axial chirality because they form stable rotational isomers, or atropisomers, that are non-superimposable mirror images of each other. Although chiral PCBs undergo atropselective biotransformation and atropselectively alter biological processes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service