Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

512737

Sigma-Aldrich

3-Methylpicolinonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N2
CAS Number:
Molecular Weight:
118.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

83-87 °C (lit.)

SMILES string

Cc1cccnc1C#N

InChI

1S/C7H6N2/c1-6-3-2-4-9-7(6)5-8/h2-4H,1H3

InChI key

WBXZCDIZXWDPBL-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient Mo (VI)-Catalyzed Hydration of Nitrile with Acetaldoxime.
Ma X, et al.
Synthetic Communications, 44(4), 474-480 (2014)
Jonita Stankevičiūtė et al.
Scientific reports, 6, 39129-39129 (2016-12-17)
Pyridinols and pyridinamines are important intermediates with many applications in chemical industry. The pyridine derivatives are in great demand as synthons for pharmaceutical products. Moreover, pyridines are used either as biologically active substances or as building blocks for polymers with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service