Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

511579

Sigma-Aldrich

Tetrakis(triphenylphosphine)palladium, polymer-bound

100-200 mesh, extent of labeling: 0.4-0.9 mmol/g loading, 2 % cross-linked with divinylbenzene

Synonym(s):

Palladium-tetrakis(triphenylphosphine), polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

solid

crosslinking

2 % cross-linked with divinylbenzene

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reaction type: solution phase peptide synthesis
reagent type: catalyst

extent of labeling

0.4-0.9 mmol/g loading

particle size

100-200 mesh

storage temp.

2-8°C

SMILES string

[Pd].c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9.c%10ccc(cc%10)P(c%11ccccc%11)c%12ccccc%12

InChI

1S/4C18H15P.Pd/c4*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;/h4*1-15H;

InChI key

NFHFRUOZVGFOOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Polymer-bound catalyst for Suzuki Coupling reactions.,

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jang, S-B.
Tetrahedron Letters, 38, 4421-4421 (1997)
Jang, S-B.
Tetrahedron Letters, 38, 1793-1793 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service