Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

511501

Sigma-Aldrich

Di-tert-butyl selenide

≥99.99%, electronic grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Se(C(CH3)3)2
CAS Number:
Molecular Weight:
193.19
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

electronic grade

vapor pressure

16 mmHg ( 54 °C)

Assay

≥99.99%

form

liquid

bp

123 °C (lit.)

density

1.048 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)[Se]C(C)(C)C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service