Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

511269

Sigma-Aldrich

Tris(tetramethylcyclopentadienyl)samarium(III)

99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H39Sm
CAS Number:
Molecular Weight:
513.96
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.9%

reaction suitability

core: samarium
reagent type: catalyst

SMILES string

[Sm].C[C]1[CH][C](C)[C](C)[C]1C.C[C]2[CH][C](C)[C](C)[C]2C.C[C]3[CH][C](C)[C](C)[C]3C

InChI

1S/3C9H13.Sm/c3*1-6-5-7(2)9(4)8(6)3;/h3*5H,1-4H3;

InChI key

PVGFZJIEOXUPET-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Supplementary Hazards

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service