Skip to Content
MilliporeSigma
All Photos(1)

Documents

50476

Sigma-Aldrich

Bis[4-(3,3,4,4,5,5,5-heptafluoro-2,2-bis(trifluoromethyl)pentyl)phenyl]phenylphosphine

≥90.0%

Synonym(s):

Bis[4-(2,2-bis-(trifluoromethyl)-1H,1H,2H,2H-perfluoropentyl)phenyl]phenylphosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H17F26P
CAS Number:
Molecular Weight:
926.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥90.0%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Fluorous synthesis

refractive index

n20/D 1.459

functional group

phosphine

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(Cc1ccc(cc1)P(c2ccccc2)c3ccc(CC(C(F)(F)F)(C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)F)cc3)(C(F)(F)F)C(F)(F)F

InChI

1S/C32H17F26P/c33-23(34,25(37,38)31(53,54)55)21(27(41,42)43,28(44,45)46)14-16-6-10-19(11-7-16)59(18-4-2-1-3-5-18)20-12-8-17(9-13-20)15-22(29(47,48)49,30(50,51)52)24(35,36)26(39,40)32(56,57)58/h1-13H,14-15H2

InChI key

JKFFEJFWOMHYDX-UHFFFAOYSA-N

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service